1,3-Dimethoxy-5-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-3,5-dimethoxybenzene

A159671-100G A159671-1G A159671-25G A159671-5G
77191-030EA 444.96 USD
77191-030 77191-032 77191-034 77191-036
1,3-Dimethoxy-5-fluorobenzene 98%
1,3-Dimethoxy-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
MDL Number: MFCD00012445
CAS Number: 52189-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR