Ethyl 7-bromobenzo[d][1,3]dioxole-4-carboxylate 96%

Supplier: Ambeed
Warning

A159440-100MG A159440-250MG
77163-144EA 126.69 USD
77163-144 77163-146
Ethyl 7-bromobenzo[d][1,3]dioxole-4-carboxylate 96%
Ethyl 7-bromobenzo[d][1,3]dioxole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
MDL Number: MFCD20487962
CAS Number: 1312610-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR