4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%

Supplier: Ambeed
Warning

Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol

A159402-100G A159402-25G A159402-500G A159402-5G
77191-022EA 21.63 USD
77191-022 77191-024 77191-026 77191-028
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
MDL Number: MFCD00191220
CAS Number: 2403-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR