2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Methoxymethyl)phenylboronic acid, pinacol ester

A159211-1G A159211-250MG A159211-5G
77163-138EA 80.34 USD
77163-138 77163-140 77163-142
2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
MDL Number: MFCD06795646
CAS Number: 675605-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR