1-(Carbamoylmethyl)pyridinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Amino-2-oxoethyl)pyridin-1-ium chloride

A159205-1G A159205-5G
77191-010EA 19.57 USD
77191-010 77191-012
1-(Carbamoylmethyl)pyridinium chloride 98%
1-(Carbamoylmethyl)pyridinium chloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
MDL Number: MFCD00032002
CAS Number: 41220-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR