(S)-N-Boc-4-fluorophenylglycine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-4-F-Phg-OH, (S)-2-((tert-Butoxycarbonyl)amino)-2-(4-fluorophenyl)acetic acid

A158747-100MG A158747-250MG A158747-5G
77191-000EA 28.84 USD
77191-000 77191-002 77746-083
(S)-N-Boc-4-fluorophenylglycine 95%
(S)-N-Boc-4-fluorophenylglycine

Precaución: for Research Use Only

Formula: C₁₃H₁₆FNO₄
MW: 269.272 g/mol
MDL Number: MFCD01320862
CAS Number: 142186-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR