N-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]thiophen-2-yl)propionamide 95%

Supplier: Ambeed
Warning

A158729-100MG A158729-250MG
77163-108EA 13.39 USD
77163-108 77163-110
N-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]thiophen-2-yl)propionamide 95%
N-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]thiophen-2-yl)propionamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.32 g/mol
MDL Number: MFCD00992942
CAS Number: 61627-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR