4-Phenylthiophen-2-amine 95%

Supplier: Ambeed
Warning

A158605-100MG A158605-1G
77163-098EA 113 USD
77163-098 77567-304
4-Phenylthiophen-2-amine 95%
4-Phenylthiophen-2-amine

Precaución: For research use only.

Formula: C₁₀H₉NS
MW: 175.25 g/mol
MDL Number: MFCD11896263
CAS Number: 67637-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR