(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-trifluoromethoxyphenylboronic acid

A158462-1G A158462-250MG A158462-5G
77190-984EA 78 USD
77190-984 77190-986 77190-988
(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid 98%
(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₄
MW: 235.95 g/mol
MDL Number: MFCD08059517
CAS Number: 290832-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR