5-tert-Butyl 4-ethyl 3-methyl-6,7-dihydro-3H-imidazo[4,5-c]pyridine-4,5(4H)-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
A158243-100MG
A158243-250MG
77163-094EA
133.9
USD
77163-094
77163-096
5-tert-Butyl 4-ethyl 3-methyl-6,7-dihydro-3H-imidazo[4,5-c]pyridine-4,5(4H)-dicarboxylate 95%
5-tert-Butyl 4-ethyl 3-methyl-6,7-dihydro-3H-imidazo[4,5-c]pyridine-4,5(4H)-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₃N₃O₄ MW: 309.36 g/mol |
MDL Number:
MFCD22393526 CAS Number: 1380679-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...