4-Bromo-1,3-dihydro-2-benzofuran 95%

Supplier: Ambeed
Warning

A158174-1G A158174-250MG A158174-5G A158174-100MG
77163-084EA 69 USD
77163-084 77163-086 77163-088 77892-082
4-Bromo-1,3-dihydro-2-benzofuran 95%
4-Bromo-1,3-dihydro-2-benzofuran

Precaución: for Research Use Only

Formula: C₈H₇BrO
MDL Number: MFCD22397733
CAS Number: 1402667-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR