Methyl 4-nitro-2-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A157360-100MG A157360-1G A157360-250MG A157360-5G
77163-062EA 9.27 USD
77163-062 77163-064 77163-066 77611-504
Methyl 4-nitro-2-(trifluoromethyl)benzoate 97%
Methyl 4-nitro-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₄
MW: 249.143 g/mol
MDL Number: MFCD14698082
CAS Number: 900254-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR