Boc-N-methyl-DL-alanine 97%
Supplier: Ambeed
|
Warning
|
A157307-1G
A157307-250MG
A157307-25G
A157307-5G
A157307-100G
77163-048EA
17
USD
77163-048
77163-050
77163-052
77163-054
77892-077
Boc-N-methyl-DL-alanine 97%
Boc-N-methyl-DL-alanine
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₄ |
MDL Number:
MFCD00153297 CAS Number: 13734-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...