(R)-2-Methoxypropan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A157239-100MG A157239-1G A157239-250MG A157239-5G
77163-038EA 46.35 USD
77163-038 77163-040 77163-042 77683-313
(R)-2-Methoxypropan-1-amine hydrochloride 95%
(R)-2-Methoxypropan-1-amine hydrochloride

Precaución: For research use only.

Formula: C₄H₁₂ClNO
MW: 125.60 g/mol
MDL Number: MFCD18382032
CAS Number: 907545-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR