1,5-Dibromo-3-methylpentane 98%

Supplier: Ambeed
Warning

A157201-1G A157201-25G A157201-5G A157201-100G
77190-970EA 11 USD
77190-970 77190-972 77190-974 77615-280
1,5-Dibromo-3-methylpentane 98%
1,5-Dibromo-3-methylpentane

Precaución: for Research Use Only

Formula: C₆H₁₂Br₂
MDL Number: MFCD00039200
CAS Number: 4457-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR