PFI-4 98%

Supplier: Ambeed
Warning

Synonyms: N-(1,3-Dimethyl-2-oxo-6-(pyrrolidin-1-yl)-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methoxybenzamide, N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide

A157158-100MG A157158-10MG A157158-1MG A157158-50MG A157158-5MG
77163-016EA 243.08 USD
77163-016 77163-018 77163-020 77163-022 77163-024
PFI-4 98%
PFI-4

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₄O₃
MW: 380.44 g/mol
MDL Number: MFCD14744412
CAS Number: 900305-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR