3,4-Thiophenedicarboxylic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: Thieno[3,4-c]furan-1,3-dione

A157089-100MG A157089-1G A157089-250MG
77163-004EA 11 USD
77163-004 77163-006 77163-008
3,4-Thiophenedicarboxylic anhydride 95%
3,4-Thiophenedicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₆H₂O₃S
MDL Number: MFCD27976700
CAS Number: 6007-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR