7,8-Dihydro-5(6H)-quinolinone 97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
7,8-Dihydroquinolin-5(6H)-one
A156644-100MG
A156644-10G
A156644-250MG
A156644-25G
A156644-5G
A156644-1G
77162-982EA
7.21
USD
77162-982
77162-984
77162-986
77162-988
77162-990
77683-311
7,8-Dihydro-5(6H)-quinolinone 97%, Ambeed.Inc
7,8-Dihydro-5(6H)-quinolinone
Precaución: For research use only.
Formula:
C₉H₉NO MW: 147.18 g/mol |
MDL Number:
MFCD02181161 CAS Number: 53400-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...