Diethyl 3-oxocyclopentane-1,1-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
A156531-250MG
A156531-1G
77190-962EA
98
USD
77190-962
77746-073
Diethyl 3-oxocyclopentane-1,1-dicarboxylate 97%
Diethyl 3-oxocyclopentane-1,1-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₆O₅ |
MDL Number:
MFCD28155104 CAS Number: 180573-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...