3-(Methylamino)benzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A156517-100MG A156517-1G A156517-250MG
77162-974EA 24.72 USD
77162-974 77162-976 77162-978
3-(Methylamino)benzoic acid hydrochloride 97%
3-(Methylamino)benzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉NO₂·ClH
MW: 187.624 g/mol
MDL Number: MFCD09752165
CAS Number: 1194804-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR