2,5-Dimethyl-p-phenylenediamine 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Diamino-p-xylene, 2,5-Dimethyl-1,4-phenylenediamine, 1,4-Diamino-2,5-dimethylbenzene, 2,5-Dimethylbenzene-1,4-diamine

A156290-100G A156290-25G A156290-500G A156290-5G
77190-948EA 59 USD
77190-948 77190-950 77190-952 77190-954
2,5-Dimethyl-p-phenylenediamine 95%
2,5-Dimethyl-p-phenylenediamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00052675
CAS Number: 6393-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR