Copper iodide 99%

Supplier: Ambeed
Warning

A156258-25G A156258-500G A156258-5G A156258-100G A156258-1000G
77162-962EA 13 USD
77162-962 77162-964 77162-966 77549-982 77611-488
Copper iodide 99%
Copper iodide

Precaución: for Research Use Only

Formula: CuI
MDL Number: MFCD00010978
CAS Number: 1335-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR