[2,2'-Bipyridine]-5-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A156017-100MG
A156017-250MG
77162-946EA
56.65
USD
77162-946
77162-948
[2,2'-Bipyridine]-5-carbaldehyde 95%
[2,2'-Bipyridine]-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₈N₂O MW: 184.19 g/mol |
MDL Number:
MFCD11042460 CAS Number: 179873-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...