(E)-1-Chloro-4-(2-nitrovinyl)benzene 97%

Supplier: Ambeed
Danger

A155974-1G A155974-25G A155974-5G
77190-936EA 13.39 USD
77190-936 77190-938 77190-940
(E)-1-Chloro-4-(2-nitrovinyl)benzene 97%
(E)-1-Chloro-4-(2-nitrovinyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆ClNO₂
MW: 183.59 g/mol
MDL Number: MFCD00024825
CAS Number: 5153-70-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR