tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-methyl-4-oxopiperidine-1-carboxylate
A155859-100G
A155859-10G
A155859-1G
A155859-250MG
A155859-25G
A155859-5G
77190-924EA
420.24
USD
77190-924
77190-926
77190-928
77190-930
77190-932
77190-934
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 97%
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₃ MW: 213.2735 g/mol |
MDL Number:
MFCD08460962 CAS Number: 181269-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...