6-((6-(1-Methyl-1H-pyrazol-4-yl)imidazo[1,2-b]pyridazin-3-yl)methyl)quinoline ≥98%

Supplier: Ambeed
Warning

A155854-10MG A155854-1MG A155854-50MG A155854-5MG
77190-916EA 72 USD
77190-916 77190-918 77190-920 77190-922
6-((6-(1-Methyl-1H-pyrazol-4-yl)imidazo[1,2-b]pyridazin-3-yl)methyl)quinoline ≥98%
6-((6-(1-Methyl-1H-pyrazol-4-yl)imidazo[1,2-b]pyridazin-3-yl)methyl)quinoline

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₆
MDL Number: MFCD22420820
CAS Number: 1185763-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR