4-(Methylthio)aniline 97%

Supplier: Ambeed
Danger

A155124-100G A155124-25G A155124-5G A155124-10G
77190-898EA 51.01 USD
77190-898 77190-900 77190-902 77746-067
4-(Methylthio)aniline 97%
4-(Methylthio)aniline

Precaución: for Research Use Only

Formula: C₇H₉NS
MDL Number: MFCD00007889
CAS Number: 104-96-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR