2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

A155113-1G A155113-250MG
77190-894EA 58.71 USD
77190-894 77190-896
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide 95%
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂OS
MW: 182.25 g/mol
MDL Number: MFCD00122463
CAS Number: 77651-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR