Ethyl 2-((5-chloropyridin-2-yl)amino)-2-oxoacetate hydrochloride 97%

Supplier: Ambeed
Danger

A155033-10G A155033-5G A155033-100G
77162-930EA 16.48 USD
77162-930 77162-932 77683-301
Ethyl 2-((5-chloropyridin-2-yl)amino)-2-oxoacetate hydrochloride 97%
Ethyl 2-((5-chloropyridin-2-yl)amino)-2-oxoacetate hydrochloride

Precaución: For research use only.

Formula: C₉H₁₀Cl₂N₂O₃
MW: 265.09 g/mol
MDL Number: MFCD02625495
CAS Number: 1243308-37-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR