3-Acetyl-2-bromopyridine 97%

Supplier: Ambeed
Warning

A154173-1G A154173-250MG A154173-5G A154173-100MG
77162-916EA 55 USD
77162-916 77162-918 77162-920 77892-053
3-Acetyl-2-bromopyridine 97%
3-Acetyl-2-bromopyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MDL Number: MFCD04114370
CAS Number: 84199-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR