tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A153934-100MG A153934-250MG
77162-900EA 217 USD
77162-900 77162-902
tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate 98%
tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MDL Number: MFCD15474948
CAS Number: 1260639-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR