(1R,3R)-3-Aminocyclopentanecarboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

A153921-100MG A153921-1G A153921-250MG
77162-894EA 49 USD
77162-894 77162-896 77162-898
(1R,3R)-3-Aminocyclopentanecarboxylic acid hydrochloride 98%
(1R,3R)-3-Aminocyclopentanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD27989239
CAS Number: 1625682-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR