1,2-Bis(triethoxysilyl)ethane 95%

Supplier: Ambeed
Warning

Synonyms: Ethylenebis(triethoxysilane), 4,4,7,7-Tetraethoxy-3,8-dioxa-4,7-disiladecane

A153151-1000G A153151-100G A153151-10G A153151-25G A153151-500G
77190-860EA 109.18 USD
77190-860 77190-862 77190-864 77190-866 77190-868
1,2-Bis(triethoxysilyl)ethane 95%
1,2-Bis(triethoxysilyl)ethane

Precaución: for Research Use Only

Formula: C₁₄H₃₄O₆Si₂
MW: 354.59 g/mol
MDL Number: MFCD00239362
CAS Number: 16068-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR