2-(2-isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid 2,2,2-Trifluoroacetic acid salt 97%

Supplier: Ambeed
Warning

A153147-100MG A153147-250MG
77162-884EA 51.5 USD
77162-884 77162-886
2-(2-isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid 2,2,2-Trifluoroacetic acid salt 97%
2-(2-isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid 2,2,2-Trifluoroacetic acid salt

Precaución: for Research Use Only

Formula: C₁₃H₁₄F₃N₅O₆
MW: 393.28 g/mol
MDL Number: MFCD28144962
CAS Number: 1638487-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR