2-(3,5-Dichloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A153092-1G A153092-250MG A153092-5G
77190-854EA 113 USD
77190-854 77190-856 77190-858
2-(3,5-Dichloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3,5-Dichloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BCl₂O₃
MDL Number: MFCD12407203
CAS Number: 942069-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR