1-(4-Amino-3-bromophenyl)ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-bromoacetophenone

A152633-10G A152633-1G A152633-25G A152633-5G
77190-836EA 118.45 USD
77190-836 77190-838 77190-840 77190-842
1-(4-Amino-3-bromophenyl)ethanone 95%
1-(4-Amino-3-bromophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.07 g/mol
MDL Number: MFCD00272386
CAS Number: 56759-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR