Adenine sulphate dihydrate 95+%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopurine sulphate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate

A152603-100G A152603-25G A152603-500G
77162-872EA 98 USD
77162-872 77162-874 77162-876
Adenine sulphate dihydrate 95+%
Adenine sulphate dihydrate

Precaución: for Research Use Only

Formula: (C₅H₅N₅)₂·H₂SO₄·2H₂O
MW: 404.37 g/mol
MDL Number: MFCD00213655
CAS Number: 6509-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR