1-(6-Ethoxypyridin-3-yl)ethanone 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A152572-5G A152572-1G
77162-868EA 119.48 USD
77162-868 77683-281
1-(6-Ethoxypyridin-3-yl)ethanone 95%, Ambeed.Inc
1-(6-Ethoxypyridin-3-yl)ethanone

Precaución: For research use only.

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
MDL Number: MFCD11553454
CAS Number: 885229-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR