3-Acetyl-2,5-dimethylthiophene 98%

Supplier: Ambeed
Warning

A152459-100G A152459-1G A152459-25G A152459-5G
77190-826EA 385.22 USD
77190-826 77190-828 77190-830 77190-832
3-Acetyl-2,5-dimethylthiophene 98%
3-Acetyl-2,5-dimethylthiophene

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MW: 154.23 g/mol
MDL Number: MFCD00009763
CAS Number: 2530-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR