4-Bromo-α-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol

A151858-100G A151858-1G A151858-25G A151858-5G
77190-816EA 169.95 USD
77190-816 77190-818 77190-820 77190-822
4-Bromo-α-methylbenzyl alcohol 98%
4-Bromo-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD00004514
CAS Number: 5391-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR