N-(3-Acetyl-5-(benzyloxy)-2-hydroxyphenyl)-2-chloroacetamide 97%

Supplier: Ambeed
Warning

A151644-100MG A151644-1G A151644-250MG
77162-842EA 31.93 USD
77162-842 77162-844 77162-846
N-(3-Acetyl-5-(benzyloxy)-2-hydroxyphenyl)-2-chloroacetamide 97%
N-(3-Acetyl-5-(benzyloxy)-2-hydroxyphenyl)-2-chloroacetamide

Precaución: for Research Use Only

Formula: C₁₇H₁₆ClNO₄
MW: 333.77 g/mol
MDL Number: MFCD28902468
CAS Number: 1799412-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR