2-(Diphenylphosphino)acetic acid 90%

Supplier: Ambeed
Warning

A151185-100MG A151185-1G A151185-250MG
77162-826EA 58 USD
77162-826 77162-828 77162-830
2-(Diphenylphosphino)acetic acid 90%
2-(Diphenylphosphino)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃O₂P
MDL Number: MFCD00443983
CAS Number: 3064-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR