tert-Butyl (1-acetylpiperidin-4-yl)carbamate 97%

Supplier: Ambeed
Warning

A151047-1G A151047-5G
77162-798EA 9 USD
77162-798 77162-800 77162-802
tert-Butyl (1-acetylpiperidin-4-yl)carbamate 97%
tert-Butyl (1-acetylpiperidin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₃
MDL Number: MFCD11844779
CAS Number: 283167-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR