Ledipasvir ≥98%

Supplier: Ambeed
Warning

Synonyms: GS-5885, Harvoni, Methyl N-[(2S)-1-[(6S)-6-[5-[9,9-Difluoro-7-[2-[(1S,2S,4R)-3-[(2S)-2-(methoxycarbonylamino)-3-methylbutanoyl]-3-azabicyclo[2.2.1]heptan-2-yl]-3H-benzimidazol-5-yl]fluoren-2-yl]-1H-imidazol-2-yl]-5-azaspiro[2.4]heptan-5-yl]-3-methyl-1-oxobu, Methyl ((S)-1-((S)-6-(5-(9,9-difluoro-7-(2-((1R,3S,4S)-2-((methoxycarbonyl)-L-valyl)-2-azabicyclo[2.2.1]heptan-3-yl)-1H-benzo[d]imidazol-6-yl)-9H-fluoren-2-yl)-1H-imidazol-2-yl)-5-azaspiro[2.4]heptan-5-yl)-3-methyl-1-oxobutan-2-yl)carbamate

A150793-10MG A150793-1G A150793-25MG A150793-5MG
77162-784EA 6.18 USD
77162-784 77162-786 77162-788 77162-790
Ledipasvir ≥98%
Ledipasvir

Precaución: for Research Use Only

Formula: C₄₉H₅₄F₂N₈O₆
MW: 889.0 g/mol
MDL Number: MFCD25976756
CAS Number: 1256388-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR