3-Amino-6-fluoro-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1H-indazol-3-amine

A150713-10G A150713-1G A150713-250MG A150713-5G
77162-776EA 303.85 USD
77162-776 77162-778 77162-780 77162-782
3-Amino-6-fluoro-1H-indazole 97%
3-Amino-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.1400 g/mol
MDL Number: MFCD06739129
CAS Number: 404827-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR