N-Boc-D-phenylalaninol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phenylalaninol

A150503-100G A150503-10G A150503-1G A150503-25G A150503-5G
77190-778EA 75 USD
77190-778 77190-780 77190-782 77190-784 77190-786
N-Boc-D-phenylalaninol 97%
N-Boc-D-phenylalaninol

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MDL Number: MFCD00216472
CAS Number: 106454-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR