Methyl 2-cyano-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

A150487-250MG A150487-5G A150487-1G
77162-758EA 8 USD
77162-758 77549-894 77892-027
Methyl 2-cyano-3-nitrobenzoate 97%
Methyl 2-cyano-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MDL Number: MFCD25967115
CAS Number: 77326-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR