4-Methoxy-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A150485-100MG A150485-250MG A150485-1G A150485-5G
77162-752EA 24.72 USD
77162-752 77162-754 77683-272 77683-273
4-Methoxy-3-pyridineboronic acid pinacol ester 98%
4-Methoxy-3-pyridineboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₂H₁₈BNO₃
MW: 235.0872 g/mol
MDL Number: MFCD07367561
CAS Number: 758699-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR