DAPT 98%

Supplier: Ambeed
Warning

Synonyms: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester

A150458-10MG A150458-1MG A150458-5MG A150458-100MG A150458-1G A150458-250MG
77162-746EA 31.93 USD
77162-746 77162-748 77162-750 77683-269 77683-270 77683-271
DAPT 98%
DAPT

Precaución: For research use only.

Formula: C₂₃H₂₆F₂N₂O₄
MW: 432.47 g/mol
MDL Number: MFCD04974585
CAS Number: 208255-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR