Ethyl 6-iodonicotinate 95%

Supplier: Ambeed
Warning

A150398-1G A150398-250MG A150398-5G
77190-772EA 114.33 USD
77190-772 77190-774 77190-776
Ethyl 6-iodonicotinate 95%
Ethyl 6-iodonicotinate

Precaución: for Research Use Only

Formula: C₈H₈INO₂
MW: 277.05 g/mol
MDL Number: MFCD16556298
CAS Number: 151917-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR